• 主页
  • 查找结果
  • Copper and nickel partitioning with nanoscale goethite under variable aquatic conditions.
Copper and nickel partitioning with nanoscale goethite under variable aquatic conditions.
Environmental toxicology and chemistry (2015-03-12)
Kelsey M Danner, Chad R Hammerschmidt, David M Costello, G Allen Burton
摘要
Metal contaminated sediments can be toxic to aquatic organisms and are common in human-dominated ecosystems, which results in metals being a leading cause of ecosystem impairment. Bioavailability of metals is influenced by their affinity for dissolved and solid-phase ligands, including iron (Fe) oxyhydroxides, which have been hypothesized to reduce metal toxicity in sediments. The authors examined the adsorption kinetics of copper (Cu) and nickel (Ni) with goethite (α-FeOOH) and characterized the influences of solute metal concentration, pH, ionic strength, and humate concentration on steady-state partitioning of the metals with goethite under conditions representative of natural aquatic environments. Copper and Ni readily adsorbed to goethite, and steady-state partitioning was achieved within 2 h. Although ionic strength had no effect on metal partitioning, adsorption of Cu and Ni to goethite was enhanced by alkaline pH and reduced by competition with humate. Because distribution coefficient (KD ) values for Cu and Ni from the present study are comparable to values measured in natural systems, the authors hypothesize that goethite may contribute significantly to the adsorption of both Ni and Cu to particles in the environment. The authors suggest that incorporating binding by Fe oxides in metal bioavailability models should be a priority for improving risk assessment of metal-contaminated oxic sediments.
材料
货号
品牌
产品描述
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
盐酸 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
盐酸, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
硝酸, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
盐酸 溶液, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
硝酸, ACS reagent, ≥90.0%
Sigma-Aldrich
氯化氢, ReagentPlus®, ≥99%
Sigma-Aldrich
盐酸 溶液, 32 wt. % in H2O, FCC
Sigma-Aldrich
盐酸 溶液, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
氯化氢 溶液, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
针铁矿, 30-63% Fe
Sigma-Aldrich
氢氧化钠, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
氢氧化钠, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-乙基-2,4-戊二酮,互变异构体的混合物, 98%
Sigma-Aldrich
硝酸-14N 溶液, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
氢氧化钠-16O 溶液, 20 wt. % in H216O, 99.9 atom % 16O