Neuronal Networks on Nanocellulose Scaffolds.
Tissue engineering. Part C, Methods (2015-09-24)
Malin Jonsson, Christian Brackmann, Maja Puchades, Karoline Brattås, Andrew Ewing, Paul Gatenholm, Annika Enejder
摘要
Proliferation, integration, and neurite extension of PC12 cells, a widely used culture model for cholinergic neurons, were studied in nanocellulose scaffolds biosynthesized by Gluconacetobacter xylinus to allow a three-dimensional (3D) extension of neurites better mimicking neuronal networks in tissue. The interaction with control scaffolds was compared with cationized nanocellulose (trimethyl ammonium betahydroxy propyl [TMAHP] cellulose) to investigate the impact of surface charges on the cell interaction mechanisms. Furthermore, coatings with extracellular matrix proteins (collagen, fibronectin, and laminin) were investigated to determine the importance of integrin-mediated cell attachment. Cell proliferation was evaluated by a cellular proliferation assay, while cell integration and neurite propagation were studied by simultaneous label-free Coherent anti-Stokes Raman Scattering and second harmonic generation microscopy, providing 3D images of PC12 cells and arrangement of nanocellulose fibrils, respectively. Cell attachment and proliferation were enhanced by TMAHP modification, but not by protein coating. Protein coating instead promoted active interaction between the cells and the scaffold, hence lateral cell migration and integration. Irrespective of surface modification, deepest cell integration measured was one to two cell layers, whereas neurites have a capacity to integrate deeper than the cell bodies in the scaffold due to their fine dimensions and amoeba-like migration pattern. Neurites with lengths of >50 μm were observed, successfully connecting individual cells and cell clusters. In conclusion, TMAHP-modified nanocellulose scaffolds promote initial cellular scaffold adhesion, which combined with additional cell-scaffold treatments enables further formation of 3D neuronal networks.
材料
货号
品牌
产品描述
Sigma-Aldrich
氢氧化钠, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
氢氧化钠, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
氢氧化钠 溶液, 50% in H2O
Sigma-Aldrich
氢氧化钠, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
链霉素 硫酸盐, powder
Sigma-Aldrich
链霉素 硫酸盐, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
氢氧化钠, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
苯乙酸, 99%
Sigma-Aldrich
氯化缩水甘油基三甲基铵, technical, ≥90% (calc. based on dry substance, AT)
Sigma-Aldrich
氢氧化钠, reagent grade, 97%, powder
Sigma-Aldrich
氢氧化钠 溶液, 5.0 M
Sigma-Aldrich
氢氧化钠, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
氢氧化钠, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
氢氧化钠, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
氢氧化钠, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
氢氧化钠, reagent grade, 97%, flakes
Sigma-Aldrich
氢氧化钠, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
氢氧化钠, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
苯乙酸, ≥99%, FCC, FG
Sigma-Aldrich
链霉素 硫酸盐, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
氢氧化钠 溶液, purum, ≥32%
Sigma-Aldrich
3-乙基-2,4-戊二酮,互变异构体的混合物, 98%
Sigma-Aldrich
氢氧化钠, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
苯乙酸, suitable for plant cell culture
Sigma-Aldrich
氢氧化钠-16O 溶液, 20 wt. % in H216O, 99.9 atom % 16O