  • Search Results
  • Identification of a high-affinity monoclonal antibody against ochratoxin A and its application in enzyme-linked immunosorbent assay.
Identification of a high-affinity monoclonal antibody against ochratoxin A and its application in enzyme-linked immunosorbent assay.
Toxicon : official journal of the International Society on Toxinology (2015-09-28)
Xian Zhang, Mengjiao Sun, Yue Kang, Hui Xie, Xin Wang, Houhui Song, Xiaoliang Li, Weihuan Fang
ABSTRACT
Ochratoxin A (OTA) is one of the most commonly occurring mycotoxins produced by some species of Aspergillus and can contaminate cereal and cereal products. A high-affinity anti-OTA monoclonal antibody (mAb) was generated from a hybridoma cell line 2D8 using splenocytes from a BALB/c mouse immunized with synthesized OTA-bovine serum albumin conjugate. The mAb 2D8 is specific with high affinity (3.75 × 10(9) L/M). An indirect competitive ELISA (ic-ELISA) was then developed using this mAb for quantitative determination of OTA in corn and feed samples. Using the optimized conditions, there was good linearity between OTA concentration and competitive inhibition (y = -0.6076x + 0.2441, R(2) = 0.9923) with the working range from 2.4 to 23.6 μg/kg, IC50 at 7.6 μg/kg and lower limit of detection at 1.4 μg/kg. The recovery rates in spiked samples were 91.2-110.3%. Of the 56 corn and feed samples, this ic-ELISA and a commercial kit both found the same 13 samples positive for OTA with good linear correlation between the two methods in OTA quantification (R(2) = 0.9706). We conclude that this ic-ELISA can be used for rapid and quantitative screening of corn and feed samples for the presence of OTA.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
N-羟基丁二酰亚胺, 98%
Sigma-Aldrich
N,N′-二环己基碳二亚胺, 99%
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
胸苷, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
3,3′,5,5′-四甲基联苯胺, ≥99%
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
胸苷, ≥99%
Sigma-Aldrich
黄曲霉毒素B1 来源于黄曲霉, from Aspergillus flavus
Sigma-Aldrich
L-赖氨酸 单盐酸盐, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3,3′,5,5′-四甲基联苯胺, ≥98% (TLC)
Sigma-Aldrich
脱氧雪腐镰刀菌烯醇
Sigma-Aldrich
L-赖氨酸 单盐酸盐, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
伏马毒素 B1 来源于串珠镰刀菌, ≥98% (HPLC)
Sigma-Aldrich
3,3′,5,5′-四甲基联苯胺, ≥98.0% (NT)
Sigma-Aldrich
二甲基亚砜 溶液, 50 wt. % in H2O
Sigma-Aldrich
玉米烯酮, fungal mycotoxin
Sigma-Aldrich
胸苷, ≥99.0% (HPLC)
Sigma-Aldrich
桔霉素, from Penicillium citrinum, ≥98% (HPLC)
Sigma-Aldrich
L-赖氨酸 单盐酸盐, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
3,3′,5,5′-四甲基联苯胺, tablet, 1 mg substrate per tablet
Sigma-Aldrich
N,N′-二环己基碳二亚胺 溶液, 1.0 M in methylene chloride