• 主页
  • 查找结果
  • Reversible adapting layer produces robust single-crystal electrocatalyst for oxygen evolution.
Reversible adapting layer produces robust single-crystal electrocatalyst for oxygen evolution.
Nature communications (2015-09-01)
Ching-Wei Tung, Ying-Ya Hsu, Yen-Ping Shen, Yixin Zheng, Ting-Shan Chan, Hwo-Shuenn Sheu, Yuan-Chung Cheng, Hao Ming Chen
摘要
Electrochemically converting water into oxygen/hydrogen gas is ideal for high-density renewable energy storage in which robust electrocatalysts for efficient oxygen evolution play crucial roles. To date, however, electrocatalysts with long-term stability have remained elusive. Here we report that single-crystal Co3O4 nanocube underlay with a thin CoO layer results in a high-performance and high-stability electrocatalyst in oxygen evolution reaction. An in situ X-ray diffraction method is developed to observe a strong correlation between the initialization of the oxygen evolution and the formation of active metal oxyhydroxide phase. The lattice of skin layer adapts to the structure of the active phase, which enables a reversible facile structural change that facilitates the chemical reactions without breaking the scaffold of the electrocatalysts. The single-crystal nanocube electrode exhibits stable, continuous oxygen evolution for >1,000 h. This robust stability is attributed to the complementary nature of defect-free single-crystal electrocatalyst and the reversible adapting layer.
材料
货号
品牌
产品描述
Sigma-Aldrich
硼氢化钠, powder, ≥98.0%
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
硼氢化钠, ReagentPlus®, 99%
Sigma-Aldrich
硼氢化钠, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
硝酸钴(II) 六水合物, 99.999% trace metals basis
Sigma-Aldrich
硼氢化钠, granular, 99.99% trace metals basis
Sigma-Aldrich
硝酸钠, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
氢氧化钾, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
硼氢化钠 溶液, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
氢氧化钠, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
氢氧化钠, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
硼氢化钠, granular, 10-40 mesh, 98%
Sigma-Aldrich
硝酸钠, BioXtra, ≥99.0%
Sigma-Aldrich
硝酸钠, 99.995% trace metals basis
Sigma-Aldrich
硫酸钠, ≥99.99% trace metals basis
Sigma-Aldrich
硫酸钠, BioXtra, ≥99.0%
Sigma-Aldrich
硼氢化钠 溶液, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
硫酸钠, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
硝酸钠, BioUltra, ≥99.0% (T)
Sigma-Aldrich
硼氢化钠, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
硼氢化钠, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
3-乙基-2,4-戊二酮,互变异构体的混合物, 98%
Sigma-Aldrich
硼氢化钠 溶液, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
硫酸钠, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
氧化铝上的氢氧化钾
Sigma-Aldrich
硼氢化钠, hydrogen-storage grade, 98%
Sigma-Aldrich
硝酸钠-14N, 99.95 atom % 14N
Sigma-Aldrich
氢氧化钠-16O 溶液, 20 wt. % in H216O, 99.9 atom % 16O