• 主页
  • 查找结果
  • Regional Bias of Intratumoral Genetic Heterogeneity of Apoptosis-Related Genes BAX, APAF1, and FLASH in Colon Cancers with High Microsatellite Instability.
Regional Bias of Intratumoral Genetic Heterogeneity of Apoptosis-Related Genes BAX, APAF1, and FLASH in Colon Cancers with High Microsatellite Instability.
Digestive diseases and sciences (2015-01-21)
Mi Ryoung Choi, Min Gwak, Nam Jin Yoo, Sug Hyung Lee
摘要
Apoptosis inactivation and intratumoral heterogeneity (ITH) are common features of cancers, including colorectal cancer (CRC). Inactivation of apoptosis prolongs cancer cell survival, and ITH may contribute to CRC progression. To examine the presence and extent of mutational ITH in the pro-apoptotic genes APAF1, BAX, and FLASH and the association of mutational ITH with pathologic parameters of CRC. The ITH of mutations in the mononucleotide repeats of APAF1, BAX and FLASH in different tumors were analyzed in 16 cases of CRC with high microsatellite instability (MSI-H) and 41 cases of CRC with stable MSI/low MSI (MSS/MSI-L) by single-strand conformation polymorphism and DNA sequencing analyses. Frameshift mutations of APAF1, BAX, and FLASH were identified in 19, 31, and 6 % of CRC with MSI-H, respectively, but also in cases of CRC with MSS/MSI-L. All but one CRC with a mutation (8/9) harbored regional ITH of the APAF1, BAX and FLASH frameshift mutations. ITH, however, was not associated with histopathologic features of CRC with MSI-H, suggesting that ITH might not be related to development of the MSI-H phenotype itself, but rather to disease progression. Our results indicate that the APAF1, BAX, and FLASH genes not only harbor frameshift mutations but also demonstrate mutational ITH, which together might play a role in the tumorigenesis of CRC with MSI-H by affecting the apoptosis of cancer cells. Our data also suggest that multiregional mutation analysis is needed for a better evaluation of the mutation status in CRC.
材料
货号
品牌
产品描述
Sigma-Aldrich
氯化镁, anhydrous, ≥98%
Sigma-Aldrich
氯化镁 溶液, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
甲酰胺, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
氯化镁, powder, <200 μm
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
氯化镁, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
甲酰胺, ACS reagent, ≥99.5%
Sigma-Aldrich
氯化镁 溶液, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
氯化镁, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
氢氧化钠, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
氢氧化钠, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
氯化镁, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
氯化镁 溶液, 0.1 M
Sigma-Aldrich
3-乙基-2,4-戊二酮,互变异构体的混合物, 98%
Sigma-Aldrich
氯化镁 溶液, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
甲酰胺 溶液, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
甲酰胺 溶液, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
氢氧化钠-16O 溶液, 20 wt. % in H216O, 99.9 atom % 16O