  • Search Results
  • Hyperleptinemia suppresses aggravation of arthritis of collagen-antibody-induced arthritis in mice.
Hyperleptinemia suppresses aggravation of arthritis of collagen-antibody-induced arthritis in mice.
Journal of orthopaedic science : official journal of the Japanese Orthopaedic Association (2015-08-22)
Tadashi Okano, Tatsuya Koike, Masahiro Tada, Yuko Sugioka, Kenji Mamoto, Kentaro Inui, Hiroyuki Okihana, Ken Ebihara, Kazuwa Nakao, Hiroaki Nakamura
ABSTRACT
Leptin is an adipocytokine produced by adipocytes and controlling body weight. It is unclear whether leptin works as a proinflammatory or an anti-inflammatory cytokine. We investigated the effects of hyperleptinemia on leptin transgenic (LepTg) mice in terms of cartilage destruction, bone destruction, joint synovitis, and serum cytokine levels by using a mouse model of collagen-antibody-induced arthritis (CAIA). CAIA was induced for female age-matched 6- to 8-week-old C57BL/6 J control mice and LepTg mice. Mice were injected intraperitoneally with 5 mg of a combination of monoclonal antibody specific for type II collagen on day 0 and 12.5 mg of lipopolysaccharide (LPS) on day 3. Clinical evaluation of arthritis was monitored for 14 days, and hind paws were examined clinically and histologically. Serum cytokine levels of interleukin (IL)-1β, IL-6, IL-10, and IL-17 and tumor necrosis factor alpha (TNF-α) were also analyzed on days 0 and 5. Moreover, THP-1 cells, which are human monocytic cell line derived from an acute monocytic leukemia patient, were cultured and differentiated into macrophages. The effects of leptin on messenger RNA (mRNA) expression of IL-6 were examined by real-time quantitative polymerase chain reaction (RT-PCR). Serum leptin concentrations were approximately ninefold higher in LepTg mice (62.0 ± 20.7 ng/ml) than in control mice (7.2 ± 0.5 ng/ml). Severity of clinical paw swelling, arthritis score, synovial hyperplasia, and cartilage damage were suppressed in LepTg mice with CAIA. Although serum cytokine levels of IL-1β, IL-17, and IL-10 and TNF-α showed no significant changes in two mice, serum levels of IL-6 in LepTg mice were suppressed at day 5. Moreover, in vitro study showed that IL-6 elevation following LPS exposure in THP-1 cells was suppressed with high leptin concentrations. Our finding suggests that hyperleptinemia suppress IL-6 responses and progression of joint inflammation. Leptin may play an anti-inflammatory role under hyperleptinemia.
Sigma-Aldrich
二乙醚, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
佛波醇12-十四酸酯13-乙酸酯, ≥99% (TLC), film or powder
Sigma-Aldrich
二乙醚, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
二乙醚, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
L-谷氨酰胺, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-谷氨酰胺, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
二乙醚, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
SAFC
L-谷氨酰胺
Supelco
苏木精
Sigma-Aldrich
苏木精, certified by the Biological Stain Commission
Sigma-Aldrich
佛波醇12-十四酸酯13-乙酸酯, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
L-谷氨酰胺, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
二乙醚, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
L-谷氨酰胺, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
二乙醚, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
二乙醚, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
L-谷氨酰胺