  • Search Results
  • Analysis of Selenium Levels in Osteosarcoma Patients and the Effects of Se-Methylselenocysteine on Osteosarcoma Cells In Vitro.
Analysis of Selenium Levels in Osteosarcoma Patients and the Effects of Se-Methylselenocysteine on Osteosarcoma Cells In Vitro.
Nutrition and cancer (2015-06-30)
Gang Huang, Bi-cheng Yong, Ming-hong Xu, Jing-chun Li, Hai-hua Guo, Jing-nan Shen
ABSTRACT
The form of selenium appears to be important for preventing cancer in humans. Here, we evaluated selenium levels in the serum and bone tissue samples from osteosarcoma patients using atomic absorption spectrometry. The in vitro effects of Se-methylselenocysteine (Se-MSC) on growth, cell cycle status, and apoptosis of osteosarcoma cells were assessed using the WST-1 assay, Hoechst 33342/propidium iodide staining, and flow cytometry, respectively. In osteosarcoma cases, the mean serum selenium levels in osteosarcoma tissue and normal bone were 0.08 mg/kg and 0.03 mg/kg, respectively (P < 0.05). Serum selenium levels in osteosarcoma and non-osteosarcoma cases were 0.09 mg/L and 0.08 mg/L, respectively (P > 0.05). Se-MSC-treated MG63 cells showed altered cellular morphology, decreased viability in a time- and dose-dependent manner, and an increase in the sub-G1 cell population. Se-MSC also downregulated Bcl-2 expression and upregulated Bax. Se-MSC inhibited the proliferation of the drug-resistant osteosarcoma cell line Saos-2/MTX300 and enhanced the inhibitory effect of pirarubicin on MG63 cells. Our data demonstrate that selenium levels are significantly higher in osteosarcoma tissue than normal bone tissue in osteosarcoma patients. The results also support the anticancer effects of Se-MSC in osteosarcoma. Further development of Se-MSC as an ancillary chemotherapy agent in osteosarcoma is warranted.
Sigma-Aldrich
水, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
水, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
水, for molecular biology, sterile filtered
Sigma-Aldrich
水, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
水, BioPerformance Certified
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
水, for cell biology, sterile ultrafiltered
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
水, PCR Reagent
Sigma-Aldrich
氯化钠, 99.999% trace metals basis
Sigma-Aldrich
氯化钠, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
氯化钠, tablet
Sigma-Aldrich
E-Toxate 水, endotoxin, free
Sigma-Aldrich
低氘水, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
氯化钠, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
水-16O, ≥99.94 atom % 16O
Sigma-Aldrich
氯化钠-35Cl, 99 atom % 35Cl