  • Search Results
  • Determination of polycyclic aromatic hydrocarbons in fractions in asphalt mixtures using liquid chromatography coupled to mass spectrometry with atmospheric pressure chemical ionization.
Determination of polycyclic aromatic hydrocarbons in fractions in asphalt mixtures using liquid chromatography coupled to mass spectrometry with atmospheric pressure chemical ionization.
Journal of separation science (2015-04-18)
Paulo Cicero Nascimento, Luciana Assis Gobo, Denise Bohrer, Leandro Machado Carvalho, Margareth Coutinho Cravo, Leni Figueiredo Mathias Leite
ABSTRACT
An analytical method using liquid chromatography coupled to mass spectrometry with atmospheric pressure chemical ionization for the determination of polycyclic aromatic hydrocarbons in asphalt fractions has been developed. The 14 compounds determined, characterized by having two or more condensed aromatic rings, are expected to be present in asphalt and are considered carcinogenic and mutagenic. The parameters of the atmospheric pressure chemical ionization interface were optimized to obtain the highest possible sensitivity for all of the compounds. The limits of detection ranged from 0.5 to 346.5 μg/L and the limits of quantification ranged from 1.7 to 1550 μg/L. The method was validated against a diesel particulate extract standard reference material (NIST SRM 1975), and the obtained concentrations agreed with the certified values. The method was applied to asphalt samples after its fractionation according to ASTM D4124 and the method of Green. The concentrations of the seven polycyclic aromatic hydrocarbons quantified in the sample ranged from 0.86 mg/kg for benzo[ghi]perylene to 98.32 mg/kg for fluorene.
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
萘, 99%
Sigma-Aldrich
苯并[a]芘, ≥96% (HPLC)
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
芘, 98%
Sigma-Aldrich
蒽, ReagentPlus®, 99%
Sigma-Aldrich
芴, 98%
Sigma-Aldrich
菲, 98%
Sigma-Aldrich
苊, 99%
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
蒽, reagent grade, 97%
Sigma-Aldrich
苯并[b]荧蒽, 98%
Sigma-Aldrich
苊烯, 99%
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
苯并[ghi]苝, 98%
Sigma-Aldrich
荧蒽, 98%
Sigma-Aldrich
萘, suitable for scintillation, ≥99%
Sigma-Aldrich
芘, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
菲, sublimed grade, ≥99.5%
Sigma-Aldrich
蒽, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
苯并[k]荧蒽, for fluorescence, ≥99%
Supelco
甲醇 溶液, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
甲醇 溶液, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
乙腈, ≥99.5%, ACS reagent
Sigma-Aldrich
芘, sublimed grade, 99%
Sigma-Aldrich
甲醇, NMR reference standard
Sigma-Aldrich
乙腈 溶液, contains 0.05 % (v/v) trifluoroacetic acid