  • Search Results
  • Significant Reduction in Helicobacter pylori Load in Humans with Non-viable Lactobacillus reuteri DSM17648: A Pilot Study.
Significant Reduction in Helicobacter pylori Load in Humans with Non-viable Lactobacillus reuteri DSM17648: A Pilot Study.
Probiotics and antimicrobial proteins (2014-12-08)
Caterina Holz, Andreas Busjahn, Heidrun Mehling, Stefanie Arya, Mewes Boettner, Hajar Habibi, Christine Lang
ABSTRACT
Reducing the amount of Helicobacter pylori in the stomach by selective bacterial-bacterial cell interaction was sought as an effective and novel method for combating the stomach pathogen. Lactobacillus reuteri DSM17648 was identified as a highly specific binding antagonist to H. pylori among more than 700 wild-type strains of Lactobacillus species. Applying a stringent screening procedure, the strain DSM17648 was identified as selective binder to H. pylori cells under in vivo gastric conditions. The strain DSM17648 co-aggregates the pathogen in vivo and in vitro. The specific co-aggregation occurs between Lact. reuteri DSM17648 and different H. pylori strains and serotypes, as well as H. heilmannii, but not with Campylobacter jejuni or other commensal oral and intestinal bacteria. Lact. reuteri DSM17648 was shown in a proof-of-concept single-blinded, randomized, placebo-controlled pilot study to significantly reduce the load of H. pylori in healthy yet infected adults. Reducing the amount of H. pylori in the stomach by selective bacterial-bacterial cell interaction might be an effective and novel method for combating the stomach pathogen. Lact. reuteri DSM17648 might prove useful as an adhesion blocker in antibiotic-free H. pylori therapies.
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
蔗糖, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
蔗糖, ≥99.5% (GC)
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
蔗糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
蔗糖, BioUltra, for molecular biology, ≥99.5% (HPLC)
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
蔗糖, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
蔗糖, ACS reagent
Sigma-Aldrich
蔗糖, ≥99.5%
Sigma-Aldrich
蔗糖, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
蔗糖, Grade II, suitable for plant cell culture
Sigma-Aldrich
蔗糖, meets USP testing specifications
Sigma-Aldrich
氯化钠, 99.999% trace metals basis
Sigma-Aldrich
蔗糖, Grade I, suitable for plant cell culture
Sigma-Aldrich
氯化钠, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
氢碘酸, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
氯化钠, tablet
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
氯化钠, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
氯化钠, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
氯化钠-35Cl, 99 atom % 35Cl