  • Search Results
  • The transcription factor NR4A1 is essential for the development of a novel macrophage subset in the thymus.
The transcription factor NR4A1 is essential for the development of a novel macrophage subset in the thymus.
Scientific reports (2015-06-20)
Robert Tacke, Ingo Hilgendorf, Hannah Garner, Claire Waterborg, Kiwon Park, Heba Nowyhed, Richard N Hanna, Runpei Wu, Filip K Swirski, Frederic Geissmann, Catherine C Hedrick
ABSTRACT
Tissue macrophages function to maintain homeostasis and regulate immune responses. While tissue macrophages derive from one of a small number of progenitor programs, the transcriptional requirements for site-specific macrophage subset development are more complex. We have identified a new tissue macrophage subset in the thymus and have discovered that its development is dependent on transcription factor NR4A1. Functionally, we find that NR4A1-dependent macrophages are critically important for clearance of apoptotic thymocytes. These macrophages are largely reduced or absent in mice lacking NR4A1, and Nr4a1-deficient mice have impaired thymocyte engulfment and clearance. Thus, NR4A1 functions as a master transcription factor for the development of this novel thymus-specific macrophage subset.
Sigma-Aldrich
水, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
水, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
地塞米松, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
水, for molecular biology, sterile filtered
Sigma-Aldrich
地塞米松, ≥98% (HPLC), powder
Sigma-Aldrich
水, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
水, BioPerformance Certified
Sigma-Aldrich
水, for cell biology, sterile ultrafiltered
Sigma-Aldrich
水, PCR Reagent
Sigma-Aldrich
地塞米松, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
偶氮二甲酸二乙酯 溶液, 40 wt. % in toluene
Sigma-Aldrich
地塞米松, meets USP testing specifications
Sigma-Aldrich
E-Toxate 水, endotoxin, free
Sigma-Aldrich
二氯甲基二乙胺 盐酸盐, 98%
Sigma-Aldrich
低氘水, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
水-16O, ≥99.94 atom % 16O