Merck
Phenolic profile and biological potential of Endopleura uchi extracts.
Asian Pacific journal of tropical medicine (2015-11-29)
Luís R Silva, Rafaela Teixeira
ABSTRACT
To improve the knowledge on the metabolite composition of Endopleura uchi bark, concerning phenolic compounds, and to evaluate some of its biological capacities for further possible exploitation in food and pharmaceutical industries associated to their health-promoting qualities. Two different extracts (infusion and hydroethanolic) were studied concerning phenolic composition and biological potential. Five compounds were determined by HPLC-DAD, being bergenin the major one. In general way, infusion presents a greater richness in these metabolites. The antioxidant, acetylcholinesterase, butyrylcholinesterase, α-glucosidase and antibacterial activity were checked by in vitro assays. A dose-dependent response was noticed against DPPH, superoxide and nitric oxide radicals, acetylcholinesterase, butyrylcholinesterase and against α-glucosidase inhibitory assay. Antibacterial capacity of both extracts was investigated against Gram-positive and Gram-negative bacteria, being more effective against the first one. The concentrations of infusion extract tested here revealed that is non-toxic for intestinal (Caco-2) cells line. The results suggest that the extracts of Endopleura uchi may be interesting for incorporating in pharmaceutical preparations for human health, since it can suppress hyperglycaemia and inhibit cholinesteases, and or as food additive due to its antiradical and antibacterial activities.
Sigma-Aldrich
纯乙醇, 200 proof, for molecular biology
Sigma-Aldrich
二氯甲烷, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
纯乙醇, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
磷酸钾二元 一元, ACS reagent, ≥99.0%
Sigma-Aldrich
二氯甲烷, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
磷酸钾二元 一元, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
纯乙醇, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
N-(1-萘基)乙二胺 二盐酸盐, ACS reagent, >98%
Sigma-Aldrich
磷酸钾二元 一元, ReagentPlus®
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
二氯甲烷, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
纯乙醇, 190 proof, for molecular biology
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
甲醇, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
纯乙醇, 200 proof, meets USP testing specifications
Sigma-Aldrich
磷酸钾二元 一元, for molecular biology, ≥98.0%
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
甲醇, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
N-(1-萘基)乙二胺 二盐酸盐, ≥98%
Sigma-Aldrich
乙醇, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
二氯甲烷, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
甲醇, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
磷酸钾二元 一元, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
二氯甲烷, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer