  • Search Results
  • Metabolomics strategy reveals therapeutical assessment of limonin on nonbacterial prostatitis.
Metabolomics strategy reveals therapeutical assessment of limonin on nonbacterial prostatitis.
Food & function (2015-08-25)
Aihua Zhang, Huiyu Wang, Hui Sun, Yue Zhang, Na An, Guangli Yan, Xiangcai Meng, Xijun Wang
ABSTRACT
Limonin has been found to possess significant anti-inflammatory properties in animal tests and with, human cells, however, its precise metabolism mechanism has not been well explored. The aim of this study was to investigate the anti-inflammatory effects of limonin in a nonbacterial prostatitis (NBP) animal model. Global metabolite profiling was performed by ultra-high-performance liquid chromatography combined with time-of-flight mass spectrometry (UPLC/ESI-TOFMS) and in conjunction with multivariate data analysis and pathway analysis which were integrated to explore differentiating metabolites and clarify the mechanism of limonin against capsaicin-induced NBP. Limonin has a potential protective function revealed by the metabolic profiling of limonin-treated rats located closer to the normal group. Twenty potential biomarker candidates and several key metabolic pathways contributing to the treatment of NBP were discovered and identified. Among the pathways, the related glycine, serine and threonine metabolism, glycerophospholipid metabolism were acutely perturbed. The changes in metabolites were restored to their base-line levels after limonin treatment, which might be through regulating the perturbed pathways to the normal state. The results indicate that changed biomarkers and pathways may provide evidence and insight into limonin action mechanisms and enable us to increase research productivity toward metabolomics in therapeutical assessment and drug discovery.
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
甲醇, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
甲醇, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
甲醇, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
甲醇, BioReagent, ≥99.93%
Sigma-Aldrich
甲醇, Absolute - Acetone free
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
乙腈, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
甲醇, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
乙腈, biotech. grade, ≥99.93%
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
乙腈, ReagentPlus®, 99%
Sigma-Aldrich
DL-亮氨酸, ≥99% (HPLC)
Supelco
甲醇 溶液, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
甲醇 溶液, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
乙腈, ≥99.5%, ACS reagent
Sigma-Aldrich
甲醇, NMR reference standard
Sigma-Aldrich
乙腈, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
乙腈 溶液, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
乙腈 溶液, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
乙腈 溶液, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈