Merck
Modified hydroxyethyl starch protects cells from oxidative damage.
Carbohydrate polymers (2015-10-03)
Sergey K Filippov, Olga Yu Sergeeva, Petr S Vlasov, Margarita S Zavyalova, Galina B Belostotskaya, Vasil M Garamus, Raisa S Khrustaleva, Petr Stepanek, Nina S Domnina
ABSTRACT
This article describes the synthesis of novel starch-antioxidant conjugates, which show great potential for biomedical applications to protect cells from oxidative damage. These conjugates were synthesized by the modification of a hydroxyethyl starch (molecular weight=200,000g/mol) with various sterically hindered phenols that differ in radical scavenging activity. They possess substantial radical scavenging activity toward a model free radical. It was found that the polymer conjugate conformation depends on the antioxidant structure and degree of substitution. We constructed the complete conformational phase behavior for the polymers with increasing degrees of substitution from small-angle neutron scattering data. It was observed that the conjugate conformation changes are the result of water shifting from a thermodynamically favorable solvent to an unfavorable one, a process that then leads to compaction of the conjugate. We selected the conjugates that possess high substitution degree but still exhibit coil conformation for biological studies. The high efficiency of the conjugates was confirmed by different in vitro (hypotonic hemolysis of erythrocytes/osmotic resistance of erythrocytes and the change of [Ca(2+)]i inside freshly isolated cardiomyocytes) and in vivo (acute hemorrhage/massive blood loss) methods.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
4-(二甲氨基)吡啶, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N′-二环己基碳二亚胺, 99%
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
二甲基亚砜 溶液, 50 wt. % in H2O
Sigma-Aldrich
N,N′-二环己基碳二亚胺 溶液, 1.0 M in methylene chloride
Sigma-Aldrich
二甲基亚砜, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
二环己基碳二亚胺 溶液, 60 wt. % in xylenes
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
4-(二甲氨基)吡啶 溶液, 0.5 M in THF