• 主页
  • 查找结果
  • Gold Nanoparticles Promote Oxidant-Mediated Activation of NF-κB and 53BP1 Recruitment-Based Adaptive Response in Human Astrocytes.
Gold Nanoparticles Promote Oxidant-Mediated Activation of NF-κB and 53BP1 Recruitment-Based Adaptive Response in Human Astrocytes.
BioMed research international (2015-07-23)
Jennifer Mytych, Anna Lewinska, Jacek Zebrowski, Maciej Wnuk
摘要
Nanogold-based materials are promising candidate tools for nanobased medicine. Nevertheless, no conclusive information on their cytotoxicity is available. In the present study, we investigated the effects of gold nanoparticles (AuNPs) on human astrocytes in vitro. Nanogold treatment in a wide range of concentrations did not result in cytotoxicity. In contrast, nanogold provoked changes in the astrocyte cell cycle and induced senescence-associated β-galactosidase activity. AuNPs promoted oxidative stress and caused activation of NF-κB pathway. After nanogold treatment, an inverse correlation between the formation of 53BP1 foci and micronuclei generation was observed. The robust 53BP1 recruitment resulted in reduced micronuclei production. Thus, nanogold treatment stimulated an adaptive response in a human astrocyte cell.
材料
货号
品牌
产品描述
Sigma-Aldrich
Gold nanoparticles, 5 nm diameter, carboxylic acid functionalized, PEG 5000 coated, dispersion in H2O