  • Search Results
  • Maternal antibody transfer can lead to suppression of humoral immunity in developing zebra finches (Taeniopygia guttata).
Maternal antibody transfer can lead to suppression of humoral immunity in developing zebra finches (Taeniopygia guttata).
Physiological and biochemical zoology : PBZ (2014-09-23)
Loren Merrill, Jennifer L Grindstaff
ABSTRACT
Maternally transferred antibodies have been documented in a wide range of taxa and are thought to adaptively provide protection against parasites and pathogens while the offspring immune system is developing. In most birds, transfer occurs when females deposit immunoglobulin Y into the egg yolk, and it is proportional to the amount in the female's plasma. Maternal antibodies can provide short-term passive protection as well as specific and nonspecific immunological priming, but high levels of maternal antibody can result in suppression of the offspring's humoral immune response. We injected adult female zebra finches (Taeniopygia guttata) with one of two antigens (lipopolysaccharide [LPS] or keyhole limpet hemocyanin [KLH]) or a control and then injected offspring with LPS, KLH, or a control on days 5 and 28 posthatch to examine the impact of maternally transferred antibodies on the ontogeny of the offspring's humoral immune system. We found that offspring of females exposed to KLH had elevated levels of KLH-reactive antibody over the first 17-28 days posthatch but reduced KLH-specific antibody production between days 28 and 36. We also found that offspring exposed to either LPS or KLH exhibited reduced total antibody levels, compared to offspring that received a control injection. These results indicate that high levels of maternal antibodies or antigen exposure during development can have negative repercussions on short-term antibody production and may have long-term fitness repercussions for the offspring.
Sigma-Aldrich
磷酸盐缓冲盐水, BioPerformance Certified, pH 7.4
Sigma-Aldrich
脂多糖 来源于肠沙门氏菌 肠道血清型, purified by trichloroacetic acid extraction
Sigma-Aldrich
Anti-Chicken IgY (IgG) (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution