Merck
  • 主页
  • 查找结果
  • Mechanism of Fibronectin Binding to Human Trabecular Meshwork Exosomes and Its Modulation by Dexamethasone.
Mechanism of Fibronectin Binding to Human Trabecular Meshwork Exosomes and Its Modulation by Dexamethasone.
PloS one (2016-10-27)
W Michael Dismuke, Mikael Klingeborn, W Daniel Stamer
摘要
Exosomes are emerging as important mediators of cell-matrix interactions by means of specific adhesion proteins. Changes in the tissue-specific exosomal protein expression may underlie pathological conditions whereby extracellular matrix turnover and homeostasis is disrupted. Ocular hypertension due to extracellular matrix accumulation in the trabecular meshwork is a hallmark of glucocorticoid-induced glaucoma. In the trabecular meshwork, exosomal fibronectin mediates cell matrix interactions at cellular structures called "invadosomes". Trabecular meshwork cells use invadosomes to turn over their surrounding matrix and maintain passageways for flow of aqueous humor. In this study, we observed that human trabecular meshwork explants treated with dexamethasone released exosomes with significantly reduced amounts of fibronectin bound per exosome. Further, we found that exosome-fibronectin binding is heparan sulfate-dependent, consistent with our observation that trabecular meshwork exosomes are enriched in the heparin/heparan sulfate binding annexins A2 and A6. In this way, dexamethasone-treated explants released exosomes with a significant reduction in annexin A2 and A6 per exosome. Interestingly, we did not detect exosomal matrix metalloproteinases, but we identified abundant dipeptidyl peptidase 4, a serine protease whose activity was reduced on exosomes isolated from dexamethasone-treated explants. Together, our findings demonstrate mechanistically how corticosteroid-induced alterations in exosomal adhesion cargo and properties can account for the pathological matrix accumulation seen in many glaucoma patients.
材料
货号
品牌
产品描述
Sigma-Aldrich
OptiPrep密度梯度培养基, used for cell and subcellular organelle isolation
Sigma-Aldrich
人血浆纤维连蛋白, liquid, 0.1% (Solution), BioReagent, suitable for cell culture
Sigma-Aldrich
硫酸乙酰肝素 钠盐 来源于牛肾
Sigma-Aldrich
肝素酶II 来源于肝素黄杆菌, Lyophilized powder stabilized with approx. 25% bovine serum albumin, lyophilized powder, ≥100 units/mg protein (enzyme + BSA)