  • Search Results
  • Determination of biogenic amines in canned fish samples using head-space solid phase microextraction based on nanostructured polypyrrole fiber coupled to modified ionization region ion mobility spectrometry.
Determination of biogenic amines in canned fish samples using head-space solid phase microextraction based on nanostructured polypyrrole fiber coupled to modified ionization region ion mobility spectrometry.
Journal of chromatography. A (2016-12-26)
Razieh Parchami, Mahdie Kamalabadi, Naader Alizadeh
ABSTRACT
The head-space solid phase microextraction (HS-SPME) was applied to extraction and determination of histamine (HIS), putrescine (PUT), cadaverine (CAD), tyramine (TYR) in canned fish samples by ion mobility spectrometry (IMS) without any derivatization process. HIS and CAD have the same mobilities in nitrogen as buffer gas and their corresponding peaks are severely overlapped in ion mobility spectrum. Peak separation was acquired in the presence of 18-crown-6 vapor as complexation reagent into carrier gas and modified ionization region of IMS (MIR-IMS) at optimum flow rate. The interaction between 18-crown-6 and the mentioned amines forms nanocluster product ions with different cross section areas and ion mobilities. The effects of main extraction parameters on the efficiency of HS-SPME-MIR-IMS were investigated and optimized. Relative standard deviations (RSD%) of the biogenic amines determination at 50μgL
Product Description
Supelco
固相微萃取便携式现场采样器, coating PDMS
Supelco
固相微萃取便携式现场采样器, coating PDMS/DVB
Supelco
固相微萃取取样操作平台, for use with 15 mL vials
Supelco
固相微萃取取样操作平台, for use with 4 mL vials
Supelco
固相微萃取便携式现场采样器, coating CAR/PDMS
Sigma-Aldrich
11-(1H-吡咯-1-基)十一烷-1-硫醇, 96%
Supelco
固相微萃取 StableFlex 萃取头组合套件, for use with manual holder
Supelco
固相微萃取萃取头组合套件 1, for use with manual holder, needle size 24 ga
Supelco
固相微萃取萃取头组合套件 2, for use with manual holder, needle size 24 ga
Supelco
固相微萃取萃取头组合套件 5, needle size 23 ga, for use with autosampler
Supelco
聚丙烯酸酯 (PA) 固相微萃取萃取头, df 85 μm (PA), for use with manual holder, needle size 24 ga
Supelco
Carbowax-聚乙二醇 (PEG) 固相微萃取萃取头, needle size 23 ga, df 60 μm (PEG), metal alloy fiber, for use with manual holder
Supelco
SPME 纤维分类套件 4, for use with manual holder, size 24 ga
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME 纤维组件聚二甲基硅氧烷 (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME LC吸头,C18, tray of 96 ea
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 75 μm (CAR/PDMS), for use with manual holder, needle size 24 ga
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm (Coating thickness includes 65 μm coating + 10 μm OC (overcoating)), needle size 23 ga, pkg of 3 ea, fiber L 1 cm
Supelco
固相微萃取 StableFlex 萃取头组合套件, for use with autosampler, needle size 24 ga
Supelco
固相微萃取萃取头组合套件 1, for use with autosampler, needle size 24 ga
Supelco
固相微萃取萃取头组合套件 2, for use with autosampler, size 24 ga
Supelco
聚丙烯酸酯 (PA) 固相微萃取萃取头, df 85 μm (PA), for use with autosampler, needle size 24 ga
Supelco
Carbowax-聚乙二醇 (PEG) 固相微萃取萃取头, needle size 23 ga, df 60 μm (PEG), for use with autosampler
Supelco
SPME 纤维分类套件 4, for use with autosampler, size 24 ga
Supelco
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
SPME 纤维组件聚二甲基硅氧烷 (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME LC吸头,PDMS/DVB, pkg of 96 ea
Supelco
SPME固相微萃取萃取头 Carboxen/聚二甲基硅氧烷 (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 23 ga, for use with manual holder
Supelco
SPME固相微萃取萃取头,二甲基硅氧烷/二乙烯基苯(PDMS/DVB), df 65 μm (PDMS/DVB), needle size 24 ga, for use with manual holder
Supelco
固相微萃取 StableFlex 萃取头组合套件, needle size 23 ga, for use with autosampler