A shipment of imported garlic powder was suspected of containing peanut. Samples (subs) collected from the shipment displayed considerable variability in peanut antigenicity when analyzed by enzyme-linked immunosorbent assay (ELISA). This raised questions regarding whether peanut was actually present, the amount present, and the basis for the variability in antigenic content. Analyses that used an xMAP multiplex assay for the detection of peanut and additional food allergens generated responses that were characteristic of peanut. Specifically, the relative intensities of two different peanut-specific antibodies coupled to beads (peanut-37 and -38) and the antigen profiles were identical to garlic controls spiked with peanut. In addition, the xMAP data did not indicate the presence of other allergens. Quantitative analyses indicated an approximately fivefold variation in peanut concentration among different subs. In contrast, within a sub, the apparent peanut concentration appeared constant. Particle size analyses of the garlic powder subs indicated a single distribution profile, with a peak at 380 μm. ELISA analysis of sieve-fractionated garlic powder from one of the subs indicated that slightly less than half of the detectable peanut was smaller than 212 μm, with the remainder almost evenly split between 212 and 300 μm and >300 μm. Modeling to predict possible oral exposure levels of peanut other than those directly measured requires additional research on the physicochemical properties of peanut and garlic, along with information on the production of the garlic powder.