  • Search Results
  • Spectroscopic insight into the interaction of bovine serum albumin with imidazolium-based ionic liquids in aqueous solution.
Spectroscopic insight into the interaction of bovine serum albumin with imidazolium-based ionic liquids in aqueous solution.
Luminescence : the journal of biological and chemical luminescence (2016-11-04)
Lakkoji Satish, Sabera Millan, Harekrushna Sahoo
ABSTRACT
The study of protein-ionic liquid interactions is very important because of the widespread use of ionic liquids as protein stabilizer in the recent years. In this work, the interaction of bovine serum albumin (BSA) with different imidazolium-based ionic liquids (ILs) such as [1-ethyl-3-methyl-imidazolium ethyl sulfate (EmimESO
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)