• 主页
  • 查找结果
  • Plasmodium vivax ligand-receptor interaction: PvAMA-1 domain I contains the minimal regions for specific interaction with CD71+ reticulocytes.
Plasmodium vivax ligand-receptor interaction: PvAMA-1 domain I contains the minimal regions for specific interaction with CD71+ reticulocytes.
Scientific reports (2017-09-01)
Gabriela Arévalo-Pinzón, Maritza Bermúdez, Diana Hernández, Hernando Curtidor, Manuel Alfonso Patarroyo
摘要
The malarial parasite's invasion is complex, active and coordinated, involving many low and high affinity interactions with receptors on target cell membrane. Proteomics analysis has described around 40 proteins in P. vivax which could be involved in reticulocyte invasion; few have been studied with the aim of elucidating how many of them establish specific interactions with their respective host cells. Given the importance of knowing which of the parasite's protein regions are functionally important for invasion, minimum regions mediating specific interaction between Plasmodium vivax apical membrane antigen 1 (PvAMA-1) and its host cell were here elucidated. The region covering PvAMA-1 domains I and II (PvAMA-DI-II) specifically bound to the CD71
材料
货号
品牌
产品描述
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
胰蛋白酶 来源于牛胰腺, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
N-(二苯甲基)甲胺, ≥98.0% (GC)