• 主页
  • 查找结果
  • Largamides A-C, tiglic acid-containing cyclodepsipeptides with elastase-inhibitory activity from the marine cyanobacterium Lyngbya confervoides.
Largamides A-C, tiglic acid-containing cyclodepsipeptides with elastase-inhibitory activity from the marine cyanobacterium Lyngbya confervoides.
Planta medica (2009-02-14)
Susan Matthew, Valerie J Paul, Hendrik Luesch
摘要
Three unusual tiglic acid-containing cyclodepsipeptides, possessing the revised regioisomeric structures for largamides A-C (1-3), have been isolated from the marine cyanobacterium Lyngbya confervoides collected from southeastern Florida. The two-dimensional structures were determined by NMR spectroscopy and the absolute configurations by chiral HPLC analysis of degradation products. Compounds 1-3 are moderate inhibitors of mammalian elastase activity in vitro with IC(50) values ranging from 0.53 to 1.41 microM.
材料
货号
品牌
产品描述
Sigma-Aldrich
苯甲磺酰氟, ≥98.5% (GC)
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
反式-2-甲基-2-丁烯酸, ≥99%, FG
Sigma-Aldrich
反式-2,3-二甲基丙烯酸, 98%