• 主页
  • 查找结果
  • Deodorization of Arthrospira platensis biomass for further scale-up food applications.
Deodorization of Arthrospira platensis biomass for further scale-up food applications.
Journal of the science of food and agriculture (2017-04-22)
Sara P Cuellar-Bermúdez, Bertha Barba-Davila, Sergio O Serna-Saldivar, Roberto Parra-Saldivar, José Rodriguez-Rodriguez, Sandra Morales-Davila, Koen Goiris, Koenraad Muylaert, Cristina Chuck-Hernández
摘要
Given the importance of A. platensis as a potential food protein source, we describe an affordable deodorization process that does not significantly affect the nutritional value of algae biomass. Ethanol, acetone or hexane were used to deodorize algae biomass and then to identify the profile of volatile compounds associated with its distinctive odor. Sensorial characteristics were improved in the biomass cake after the proposed solvent extraction. Panelists identified the ethanolic extract with the most pronounced algae-related odor. Gas chromatography-mass spectrometry analysis showed that a mixture of 20 different compounds derived from fatty acids and amino acids contributed to the characteristic smell of A. platensis biomass. The results of the present study show that the ethanol solvent-free A. platensis biomass contained > 600 g kg The various extraction procedures investigated in the present study were effective in deodorizing the algae biomass. The most effective protocol was the removal of odoriferous compounds with ethanol. This particular procedure yielded an algae biomass with an improved sensorial traits. The results of the present study should help with the identification of odoriferous compounds derived from fatty acids, pigments and proteins associated with A. platensis. © 2017 Society of Chemical Industry.
材料
货号
品牌
产品描述
Sigma-Aldrich
胰蛋白酶 来源于猪胰腺, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
蛋白酶 来源于灰色链霉菌, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
聚二甲基硅氧烷, viscosity 1.0 cSt (25 °C)