• 主页
  • 查找结果
  • Bioaccessibility of four calcium sources in different whey-based dairy matrices assessed by in vitro digestion.
Bioaccessibility of four calcium sources in different whey-based dairy matrices assessed by in vitro digestion.
Food chemistry (2017-12-31)
Lucie Lorieau, Linda Le Roux, Frédéric Gaucheron, Amandine Ligneul, Etienne Hazart, Didier Dupont, Juliane Floury
摘要
Numerous calcium sources are available to enrich food, but their behavior during digestion is still unknown. This study focused on the influence of the gastro-intestinal pH, the food structure and the calcium source on the bioaccessibility of the nutrient. Four calcium sources were studied: calcium carbonate, calcium citrate malate, calcium phosphate and calcium bisglycinate. These were added to dairy matrices, containing cream and whey proteins, of different forms (liquid or gel). The kinetics of solubility and ionic calcium concentration during in vitro digestion were studied, as function of gastro-intestinal pH. All calcium sources were almost fully soluble in the gastric compartment, and then became insoluble in the intestinal phase. The level of calcium insolubilisation in the intestinal phase was not significantly influenced by the matrix structure (liquid or gel), but was more dependent on the calcium source, this effect leading to different final calcium bioaccessibility from 36% to 20%.
材料
货号
品牌
产品描述
Sigma-Aldrich
胰脂肪酶 来源于猪胰腺, 8 × USP specifications
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥2,500 units/mg protein (E1%/280)