• 主页
  • 查找结果
  • Impact of humic acid on the photoreductive degradation of perfluorooctane sulfonate (PFOS) by UV/Iodide process.
Impact of humic acid on the photoreductive degradation of perfluorooctane sulfonate (PFOS) by UV/Iodide process.
Water research (2017-10-17)
Zhuyu Sun, Chaojie Zhang, Pei Chen, Qi Zhou, Michael R Hoffmann
摘要
Iodide photolysis under UV illumination affords an effective method to produce hydrated electrons (e
材料
货号
品牌
产品描述
Sigma-Aldrich
五氟丙酸, 97%
Sigma-Aldrich
全氟戊酸, 97%
Sigma-Aldrich
全氟庚酸, ≥97.0%
Sigma-Aldrich
十三氟己烷-1-磺酸 钾盐, ≥98.0% (T)
Sigma-Aldrich
十一氟己酸, ≥97.0% (T)