Merck
99900030
Opentrons® Tube racks (4-in-1 Set)
new
Opentrons® Tube racks (4-in-1 Set)
Select a Size
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
autoclavable: no
packaging
pack of 1 ea
manufacturer/tradename
999-00030
General description
Opentrons® Tube racks (4-in-1 Set) includes 2 base stands and 4 tube holder tops: 1.5ml, 2ml, 15ml + 50ml, 15ml, and 50ml. The tube holder tops snap securely on the base stand, which fits directly on the deck. The Tube Rack Set is not autoclavable.
Legal Information
Opentrons is a registered trademark of Opentrons Labworks, Inc.