- Pseudomonas aeruginosa quorum sensing as a potential antimicrobial target.
Pseudomonas aeruginosa quorum sensing as a potential antimicrobial target.
The Journal of clinical investigation (2003-11-18)
Roger S Smith, Barbara H Iglewski
PMID14617745
ABSTRACT
Pseudomonas aeruginosa has two complete quorum-sensing systems. Both of these systems have been shown to be important for Pseudomonas virulence in multiple models of infection. Thus, these systems provide unique targets for novel antimicrobial drugs.