Skip to Content
Merck

Skip To

940526

Sodium chloride

greener alternative

Greener Alternative

anhydrous, ≥99.99% (trace metals analysis)

Synonym(s):

Common salt, Rock salt, Sodium Monochloride, Table salt, salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
UNSPSC Code:
12352200
NACRES:
NA.21
MDL number:
Assay:
≥99.99% (trace metals analysis)
Form:
crystalline powder
Solubility:
water: soluble 358 g/L at 20 °C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

High purity Salts

Quality Segment

assay

≥99.99% (trace metals analysis)

form

crystalline powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100 ppmtrace (metals basis)

pH

7

mp

801 °C

solubility

water: soluble 358 g/L at 20 °C

anion traces

nitrate (NO3-): ≤20 ppm, sulfate (SO42-): ≤20 ppm

cation traces

Al: ≤10 ppm, B: ≤10 ppm, Ba: ≤10 ppm, Ca: ≤10 ppm, Co: ≤10 ppm, Cr: ≤10 ppm, Cu: ≤10 ppm, Fe: ≤10 ppm, K: ≤10 ppm, Mn: ≤10 ppm, Ni: ≤10 ppm, Si: ≤10 ppm, Ti: ≤10 ppm, Zn: ≤10 ppm

greener alternative category

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product belongs to an Enabling category, and is crucial in molten salt mixtures for thermal energy storage in concentrating solar power systems. Its low cost and effective thermal properties enhance energy storage efficiency, supporting renewable energy use and reducing environmental impact. Click here for more information.
Sodium chloride, anhydrous is a colorless crystalline inorganic compound highly soluble in water. Our Sodium chloride anhydrous,is suitable molten salt to prepare different types of eutectic mixtures due to its high operation temperature for thermal energy storage (TES) materials and heat transfer fluids (HTFs) for next-generation concentrating solar power (CSP) systems.

Application

Molten chloride salts are considered favorable for Concentrated Solar Power (CSP) applications due to their exceptional thermal properties and high decomposition temperatures. The corrosion behavior of Inconel 625 and Hastelloy B-3 alloys was studied in a molten salt environment comprising NaCl-CaCl2-MgCl2. Other studies have documented the development of an in situ corrosion and mass transport monitoring method that utilizes a radionuclide tracing technique for studying the corrosion of 316L stainless steel (316L SS) in a NaCl–MgCl2 eutectic molten salt natural circulation loop. The eutectic salt used in this study was prepared with 58.5 mol% anhydrous sodium chloride and 41.5 mol% anhydrous magnesium chloride.
Furthermore, various MAX phases and Lewis acid salts were employed in the preparation of MXenes using the molten salt synthesis method. For example, Ti3SiC2 MAX phase and CuCl2 were mixed in a stoichiometric molar ratio of 1:3, along with sodium chloride , anhydrous and Potassium chloride, anhydrous. The mixture was heated to 750 °C and held for 24 h in an argon atmosphere. After washing with deionized water, MXene/Cu mixed particles were obtained, followed by further cleaning with APS solution, deionized water, and alcohol. The resulting MXene powders ( MS-Ti3C2Tx) were dried under vacuum at room temperature for 24 h.

Our Sodium chloride which is anhydrous, with a purity of ≥99.99%, is an excellent choice for molten salt synthesis method. Its anhydrous properties and high purity make it highly suitable for thermal energy storage (TES) materials and heat transfer fluids (HTFs) for next-generation concentrating solar power (CSP) systems and molten salt reactors (MSR).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2044397138231434
description

anhydrous, ≥99.99% (trace metals analysis)

description

99.999% trace metals basis

description

puriss. p.a., ≥99.5% (AT), powder or crystals

description

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%

form

crystalline powder

form

solid

form

powder or crystals

form

solid

assay

≥99.99% (trace metals analysis)

assay

99.999% trace metals basis

assay

≥99.5% (AT)

assay

≥99.5%

solubility

water: soluble 358 g/L at 20 °C

solubility

water: soluble (358 g/l at 20 °C (68 °F))

solubility

water: soluble 358 g/L at 20 °C

solubility

water: soluble 100 mg/mL, clear, colorless

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

impurities

≤100 ppmtrace (metals basis)

impurities

≤15.0 ppm Trace Metal Analysis

impurities

Traces of [Fe(CN)6]4- (information only), ≤0.001% total nitrogen (N)

impurities

acidity or alkalinity, complies, ≤0.0001% hexacyanoferrate(II), ≤0.001% total nitrogen (N), ≤0.005% free acid (as HCl), ≤0.005% free alkali (as NaOH), ≤0.005% in water insoluble matters, ≤0.01% Mg, alkaline earth metals (as Ca)

anion traces

nitrate (NO3-): ≤20 ppm, sulfate (SO42-): ≤20 ppm

anion traces

-

anion traces

bromide (Br-): ≤50 mg/kg, iodide (I-): ≤10 mg/kg, phosphate (PO43-): ≤5 mg/kg, sulfate (SO42-): ≤200 mg/kg

anion traces

bromide (Br-): ≤50 mg/kg, chlorate, nitrate (as NO3-): ≤30 mg/kg, iodide (I-): ≤10 mg/kg, phosphate (PO43-): ≤5 mg/kg, sulfate (SO42-): ≤10 mg/kg


Still not finding the right product?

Explore all of our products under Sodium chloride


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library