Merck
HomeNext-Generation SequencingExpert metagenomics protocols enhance metagenomics services
Expert metagenomics protocols enhance metagenomics services
Metagenomics is the analysis of the genetic composition of mixed communities of microorganisms. A metagenomics approach allows the characterization of microbial species in situ, or in the context of their naturally occurring communities. This enables the study of relevant, complex interspecies dynamics that may include metabolic complementation, lateral gene transfer, and phage-host interaction that cannot be investigated using culture-dependent methods.
AMPLICON AND WGS-BASED SEQUENCING OPTIMIZED FOR DIVERSE, LOW ABUNDANCE SAMPLES
Our team of microbiome scientists has been developing protocols for the detection of amplicon- and WGS-based (whole genome sequencing) metagenomic data acquisition. These protocols include quality controls at every step to assure low-bias results. Our experience extracting genomic information includes expert techniques for extracting nucleic acids from challenging environments like soil, or from diverse biomedical or veterinary tissues and sources including feces, saliva, mucosa, skin, and mucosal swabs.
Beyond mere bench expertise, our metagenomics service constitutes a research partnership. When next steps become unclear, our microbiome scientists extend your capabilities by consultation regarding planning, executing, analyzing your study, and interpreting your data to optimize your results.
SENSITIVE, LOW-BIAS 16S AMPLICON METAGENOMIC SEQUENCING DEVELOPED BY MICROBIOME EXPERTS
DNA from foreign organisms as internal quality controls
Lack of standardization in microbiome genomics protocols can lead to biases and error arising from sample preparation. Downstream analysis of sample amplification, sequencing and data interpretation are less reliable without internal controls. To control for such misinterpretation of metagenomic sequencing results, it is important to include a DNA standard from an organism unlikely to be found in the tested sample.
To serve as an internal control that demonstrates the accuracy and sensitivity of the 16S amplicon metagenomic sequencing service, we developed a new DNA standard from a marine bacterium Vibrio harveyi (Cat # MBD0037), which is typically not found in the human gut. Spike-in human fecal DNA samples with Vibrio harveyi at differential concentrations, showed the high accuracy and sensitivity of our 16S Amplicon metagenomic sequencing service.
DNA standard suggests high accuracy and sensitivity of the 16S Amplicon metagenomic sequencing service.
Figure 1.V harveyi DNA standart was spiked-in to stool DNA samples with differential frequencies.
EXCLUSIVE ULTRASENSITIVE 16S LIBRARY PREPARATION TECHNOLOGY: THE 5R-PLEX APPROACH
Critical biomedical and environmental samples are often plagued by the same barriers to effective metagenomic analysis:
  • Bacterial biomass in the sample is very low
  • The sample yields poor quality and highly degraded DNA
Our exclusive 5R-Plex Ultra-sensitive 16S library preparation technology is based on the SMURF (Short Multiple Regions Framework) technology which was developed to facilitate cancer microbiome studies1-3. DNA extracted from tumors in formalin-fixed and paraffin-embedded (FFPE) samples is usually highly degraded and is of low quality. Moreover, this DNA is usually characterized by low bacterial biomass and high host fraction presence, which requires an ultra-sensitive method for sequencing the 16S rRNA gene. The 5R-Plex approach is based on amplifying 5 short regions (~200bp) along the 16S rRNA gene (V2,3,5,6,8 – as illustrated in Fig. 2), followed using our exclusive algorithm to provide a larger coverage of the 16S rRNA gene. This allows better resolution of the microbial community profiling, particularly when biomass is extremely low, or DNA is likely to be highly degraded (Fig. 3).
Amplification of short regions along 16S rRNA gene to demonstrate the 5R-Plex approach.
Figure 2.5R-PLEX regions sequenced (gray) along the 16S rDNA gene
5R-plex approach suggests improved resolution of the microbial community profile.
Figure 3.The ultrasensitive 5R-plex technology was applied to two DNA samples of Sigma-Aldrich ‘Microbial community DNA mix’ (#CAT MBD0026). One was fragmented using sonication to an average fragment size of 260bp (degraded), and the other was of high molecular weight (HMW). The standard 16S (V3-V4) amplicon library prep gave an insufficient number of reads (total input) from the fragmented DNA and a poor taxonomy classification, while the 5R-PLEX method yielded high-resolution microbial profiling, even with 1pg degraded DNA.
MAKING INFORMED DECISIONS ON THE REQUIRED READ DEPTH FOR YOUR SAMPLES
Increased sequencing depth generates more reads, which enhances the sensitivity and the statistical power to detect the rarest members of a microbiome and uncover rare functional genes. Our deep shotgun sequencing (10 million reads) approach delivers the most reliable and informative microbiome description of even the most rare members of a microbiome community, such as viruses. Additionally, deep shotgun sequencing allows the deep characterization of the full functional profiling of biological pathways and strain coverage information (Fig. 4).
However, the cost of increasing read depth indiscriminately can become impractical or prohibitive. Our shallow shotgun sequencing (3 million reads) approach can provide full species-level resolution and functional profiling for large-scale studies at an affordable price.
Deep shotgun sequencing demonstrates deep characterization of the biological pathways and strain coverage.
Figure 4.Shallow sequencing with 3 million reads is sufficient to capture most taxonomic groups at the species level and the core functional pathways. For capturing the virome fraction, deep sequencing of 10 million read depth is needed.
SHOTGUN METAGENOMICS FOR LOW-INPUT DNA SAMPLES
Many biological samples have very low DNA input. Our shotgun protocol is suitable for inputs as low as 0.5ng/µl DNA input, with minimal bias. Figures 5-7 demonstrate results from an experiment in which we applied the two Shotgun library prep protocols on the same DNA samples with different inputs: 10 ng/µl for the standard protocol, and 0.5 ng/µl for the low-input protocol. The results show that there are no differences in the taxonomy profile as visualized by the stacked bar chart (Fig. 5), the alpha diversity which measures species richness and evenness within each sample at different taxonomic levels (Fig. 6) and according to the beta diversity, a measurement of microbial diversity differences between samples. The samples clusters according to the donors and not according to the library preparation protocol applied (Fig. 7).
Sample data suggests demonstrating no difference in taxonomy profiles between standard and low-input options.
Figure 5.No obvious differences in taxonomic prolific between Std vs low-input (LOWinp) Shotgun library prep protocols
Sample data suggests similar Alpha diversity between standard and low-input options.
Figure 6.Comparable Alpha diversity between Std vs low-input Shotgun library prep protocols
Sample data showing sample clusters according to the donors and not the library preparation protocol.
Figure 7.Beta-diversity clustered by donors, but not by library prep protocol (Std vs Low-input).
References
1.
Fuks G, Elgart M, Amir A, Zeisel A, Turnbaugh PJ, Soen Y, Shental N. 2018. Combining 16S rRNA gene variable regions enables high-resolution microbial community profiling. Microbiome. 6(1): https://doi.org/10.1186/s40168-017-0396-x
2.
Amir A, Zeisel A, Zuk O, Elgart M, Stern S, Shamir O, Turnbaugh PJ, Soen Y, Shental N. 2013. High-resolution microbial community reconstruction by integrating short reads from multiple 16S rRNA regions. 41(22):e205-e205. https://doi.org/10.1093/nar/gkt1070
3.
2022. The Human Tumor Microbiome Is Composed of Tumor Type–Specific Intracellular Bacteria | Science.. [Internet]. Available from: https://www.science.org/doi/10.1126/science.aay9189.