Kereta0
[Home](https://www.sigmaaldrich.com/ID/en)[Applications](https://www.sigmaaldrich.com/ID/en/applications)[Materials Science and Engineering](https://www.sigmaaldrich.com/ID/en/applications/materials-science-and-engineering)Solid State Synthesis
# Solid State Synthesis
Solid-state synthesis, or the ceramic method, is commonly used to cause a chemical reaction from solid starting materials to form a new solid with a well-defined structure. End products include polycrystalline materials, single crystals, glasses, and thin-film materials that are widely used for energy and electronic applications.
Fine-grain [metal](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/high-purity-metals-and-alloys) compounds are combined, pelletized, and heated at a controlled temperature for a specific time period. Some metal compounds, such as [metal oxides](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/oxides-and-ceramics) or [salts](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/high-purity-salts), require extreme conditions, such as high temperatures and pressure, to initiate reactions in a molten flux or a rapidly condensing vapor phase. This process is often referred to as “shake and bake” or “heat and beat” chemistry.
* * *
## Related Products
Slide 1 of 18
1 of 5
[
\
Sigma-Aldrich
\
520918
\
Gold(III) chloride trihydrate](https://www.sigmaaldrich.com/ID/id/product/aldrich/520918)
Tampilan Cepat
[
\
Sigma-Aldrich
\
554359
\
Lead(II) iodide](https://www.sigmaaldrich.com/ID/id/product/aldrich/554359)
Tampilan Cepat
[
\
Sigma-Aldrich
\
431648
\
Sodium sulfide nonahydrate](https://www.sigmaaldrich.com/ID/id/product/aldrich/431648)
Tampilan Cepat
[
\
Sigma-Aldrich
\
409308
\
Tin(II) iodide](https://www.sigmaaldrich.com/ID/id/product/aldrich/409308)
Tampilan Cepat
[
\
Sigma-Aldrich
\
229393
\
Aluminum chloride hydrate](https://www.sigmaaldrich.com/ID/id/product/aldrich/229393)
Tampilan Cepat
[
\
Sigma-Aldrich
\
206237
\
Iridium(IV) oxide](https://www.sigmaaldrich.com/ID/id/product/aldrich/206237)
Tampilan Cepat
[
\
Sigma-Aldrich
\
202908
\
Cadmium chloride](https://www.sigmaaldrich.com/ID/id/product/aldrich/202908)
Tampilan Cepat
[
\
Sigma-Aldrich
\
441899
\
Strontium](https://www.sigmaaldrich.com/ID/id/product/aldrich/441899)
Tampilan Cepat
[
\
Sigma-Aldrich
\
288497
\
Barium oxide](https://www.sigmaaldrich.com/ID/id/product/aldrich/288497)
Tampilan Cepat
[
\
Sigma-Aldrich
\
345865
\
Ammonium trifluoromethanesulfonate](https://www.sigmaaldrich.com/ID/id/product/aldrich/345865)
Tampilan Cepat
[
\
Sigma-Aldrich
\
248665
\
Calcium tungstate](https://www.sigmaaldrich.com/ID/id/product/aldrich/248665)
Tampilan Cepat
[
\
Sigma-Aldrich
\
206237
\
Iridium(IV) oxide](https://www.sigmaaldrich.com/ID/id/product/aldrich/206237)
Tampilan Cepat
[
\
Sigma-Aldrich
\
637238
\
Silicon dioxide](https://www.sigmaaldrich.com/ID/id/product/aldrich/637238)
Tampilan Cepat
[
\
Sigma-Aldrich
\
431648
\
Sodium sulfide nonahydrate](https://www.sigmaaldrich.com/ID/id/product/aldrich/431648)
Tampilan Cepat
[
\
Sigma-Aldrich
\
718467
\
Titanium(IV) oxide](https://www.sigmaaldrich.com/ID/id/product/aldrich/718467)
Tampilan Cepat
[
\
Sigma-Aldrich
\
718475
\
Aluminum oxide](https://www.sigmaaldrich.com/ID/id/product/aldrich/718475)
Tampilan Cepat
[
\
Sigma-Aldrich
\
474711
\
Barium](https://www.sigmaaldrich.com/ID/id/product/aldrich/474711)
Tampilan Cepat
[
\
Sigma-Aldrich
\
333972
\
Zirconium(IV) propoxide solution](https://www.sigmaaldrich.com/ID/id/product/aldrich/333972)
Tampilan Cepat
* * *
## Featured Categories
[](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/high-purity-salts)
[High Purity Salts](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/high-purity-salts)
We offer a wide range of high-purity salts, including nitrates, oxalates, halides, sulfates, carbonates, and acetates, available in both anhydrous and hydrated forms.
[Shop Products](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/high-purity-salts)
[](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/high-purity-metals-and-alloys)
[High-Purity Metals & Alloys](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/high-purity-metals-and-alloys)
We offer an extensive range of high-purity metals and alloys in diverse compositions and particles size, even customized for your research and production applications.
[Shop Products](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/high-purity-metals-and-alloys)
[](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/oxides-and-ceramics)
[Oxides & Ceramics](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/oxides-and-ceramics)
We offer an extensive range of high purity oxides and ceramics with specialized synthesis and purification techniques, and distinct particle sizes.
[Shop Products](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/oxides-and-ceramics)
[](https://www.sigmaaldrich.com/ID/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
[Inorganic & Metallic Nanomaterials](https://www.sigmaaldrich.com/ID/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
We offer a comprehensive portfolio of inorganic and metallic nanomaterials, functionalized nanoparticles, and nanomaterial kits for your research needs.
[Shop Products](https://www.sigmaaldrich.com/ID/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
* * *
Overview
Related Articles & Protocols
Support
The reaction rate in solid state synthesis is particularly important to characterize. Solid state reactions must go to completion, as techniques for purification of formed solids are severely limited. The rate of the solid-state reaction depends on the reaction conditions, including the structural properties, shape and surface area of the reactants, the diffusion rate, and the thermodynamic properties associated with the nucleation/reaction. The chemical and physical properties of the final materials are determined by the chemical [precursors](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/solution-and-vapor-deposition-precursors) and preparation techniques.
Modern preparation techniques for solid state are not limited to variations on the ceramic method. In solid state metathesis, reactions of metal compounds are initiated by an energy source (e.g., flame, ball mill) and propagated by the heat released during the formation of products and byproducts. [Sol-gel](https://www.sigmaaldrich.com/ID/en/products/materials-science/energy-materials/solution-and-vapor-deposition-precursors) methods utilize a concentrated or colloidal solution (the ‘sol’), which is sequentially heated, dried, and aged to form gels, coatings, and nanomaterials. Solvothermal methods involve heating solutions in a pressurized, closed vessel at temperatures above the standard boiling point of the chosen organic solvent; if the solvent is water, this is called the hydrothermal method. Many synthetic methods where a solid material is formed, such as vapor-phase depositions, intercalation, single crystal growth, and nanomaterial syntheses, can be classified as solid state synthesis.
[](https://www.sigmaaldrich.com/documents-search)
[Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search)
Visit our document search for data sheets, certificates and technical documentation.
[Find Documents](https://www.sigmaaldrich.com/documents-search)
## Related Articles
- [Solubility Rules for Ionic Compounds](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/solubility-rules-solubility-of-common-ionic-compounds)
Review solubility rules for common ionic compounds in water, including calcium carbonate, barium sulfate, and sodium sulfate, using the provided chart.
- [Using Activated Carbon as a Precious Metal Catalyst Carrier](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/activated-carbon)
Learn how activated carbon is working as a catalyst support and its advantage.
- [Magnetic Properties of Materials](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/properties)
The three types of magnetic behaviors are paramagnetism where unpaired electrons are random, ferromagnetism where unpaired electrons align, and antiferromagnetism where unpaired electrons align opposite of one another.
- [Advanced Materials for Magnetic Cooling](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/advanced-materials)
Today, near-room-temperature refrigeration is almost entirely based on a vapor-compression refrigeration cycle.
- [PCTPro-2000: Gas Sorption Analysis Tool](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/the-pctpro-2000-the-ultimate-tool-for-gas-sorption-analysis)
Gas sorption analysis aids in materials science and consumer product development.
- [See All (37)](https://www.sigmaaldrich.com/ID/en/search/facet-search?focus=sitecontent&term=facet-search)
### Find More Articles and Protocols
Enter Keywords
Search
## How Can We Help
In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/ID/en/support/customer-support)
or talk to our customer service team:
Email [custserv@sial.com](mailto:custserv@sial.com)
or call +1 (800) 244-1173
## Additional Support
- [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search)
Use the Chromatogram Search to identify unknown compounds in your sample.
- [Calculators & Apps](https://www.sigmaaldrich.com/ID/en/support/calculators-and-apps)
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- [Customer Support Request](https://www.sigmaaldrich.com/ID/en/support/customer-support)
Customer support including help with orders, products, accounts, and website technical issues.
- [FAQ](https://maestro.my.site.com/knowledgeportal/s/)
Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services.
* * *
__Featured Articles__
[Solubility Rules for Ionic Compounds](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/solubility-rules-solubility-of-common-ionic-compounds)
Review solubility rules for common ionic compounds in water, including calcium carbonate, barium sulfate, and sodium sulfate, using the provided chart.
[Using Activated Carbon as a Precious Metal Catalyst Carrier](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/activated-carbon)
Learn how activated carbon is working as a catalyst support and its advantage.
[Magnetic Properties of Materials](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/properties)
The three types of magnetic behaviors are paramagnetism where unpaired electrons are random, ferromagnetism where unpaired electrons align, and antiferromagnetism where unpaired electrons align opposite of one another.
[Advanced Materials for Magnetic Cooling](https://www.sigmaaldrich.com/ID/en/technical-documents/technical-article/materials-science-and-engineering/solid-state-synthesis/advanced-materials)
Today, near-room-temperature refrigeration is almost entirely based on a vapor-compression refrigeration cycle.
Atas
__Masuk untuk Melanjutkan__
Masuk__Tidak Punya Akun?__Daftar
Dukungan[Dukungan Pelanggan](https://www.sigmaaldrich.com/ID/id/support/customer-support)[Hubungi Kami](https://www.sigmaaldrich.com/ID/id/collections/offices)[PERTANYAAN YANG SERING DIAJUKAN](https://maestro.my.site.com/knowledgeportal/s/)[Lembar Data Keselamatan (SDS)](https://www.sigmaaldrich.com/ID/id/documents-search?tab=sds)[Sertifikat (COA/COO)](https://www.sigmaaldrich.com/ID/id/documents-search?tab=coa)[Kualitas & Regulasi](https://www.sigmaaldrich.com/ID/id/life-science/quality-and-regulatory-management)[Kalkulator & Aplikasi](https://www.sigmaaldrich.com/ID/id/support/calculators-and-apps)[Webinar](https://www.sigmaaldrich.com/ID/id/collections/webinars)
Pesanan[Pencarian Pesanan](https://www.sigmaaldrich.com/ID/id/order-lookup)[Pesanan Cepat](https://www.sigmaaldrich.com/ID/id/quick-order)[Produk Khusus](https://www.sigmaaldrich.com/ID/id/services/custom-products)[Solusi eCommerce](https://www.sigmaaldrich.com/ID/id/life-science/ecommerce/ecommerce-solutions)
Perusahaan[Tentang Kami](https://www.sigmaaldrich.com/ID/id/life-science/about-us)[Tanggung jawab](https://www.sigmaaldrich.com/ID/id/life-science/ssbi)[Acara](https://www.sigmaaldrich.com/ID/id/collections/events)[Siaran Pers](https://www.sigmaaldrich.com/ID/id/collections/press)[Program](https://www.sigmaaldrich.com/ID/id/life-science/partnership-programs)[Karir](https://careers.merckgroup.com/global/en)[Kantor](https://www.sigmaaldrich.com/ID/id/collections/offices)
Media Sosial
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
Merck
__Penelitian. Pengembangan. Produksi.__
Kami adalah pemasok terkemuka untuk industri Life Science global dengan solusi dan layanan untuk penelitian, pengembangan dan produksi bioteknologi, serta pengembangan dan produksi terapi obat farmasi.
[](https://www.sigmaaldrich.com/ID/id)
[Solusi Sigma-Aldrich® Solusi](https://www.sigmaaldrich.com/ID/id/life-science/sigma-aldrich)[Solusi BioReliance® Solusi](https://www.sigmaaldrich.com/ID/id/life-science/bioreliance)[Solusi Millipore® Solusi](https://www.sigmaaldrich.com/ID/id/life-science/millipore)[Solusi SAFC® Solusi](https://www.sigmaaldrich.com/ID/id/life-science/safc)[Solusi Milli-Q® Solusi](https://www.sigmaaldrich.com/ID/id/life-science/milliq)[Solusi Supelco® Solusi](https://www.sigmaaldrich.com/ID/id/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Jerman dan/atau afiliasinya. Dilarang memperbanyak sebagian atau seluruh isi dokumen tanpa izin tertulis dari Merck KGaA dan/atau afiliasinya, termasuk teks dan penggalian data untuk pelatihan AI dan teknologi sejenisnya.
Reproduction of any materials from the site is strictly forbidden without permission.
[Ketentuan Penggunaan Situs](https://www.sigmaaldrich.com/ID/id/life-science/legal/site-use-terms)|[Kebijakan Privasi](https://www.sigmaaldrich.com/ID/id/life-science/legal/privacy-statement)|[Syarat dan Ketentuan Umum Penjualan](https://www.sigmaaldrich.com/ID/id/life-science/legal/terms-and-conditions)|[Persetujuan Hak Cipta](https://www.sigmaaldrich.com/ID/id/life-science/legal/copyright-consent)|[Peta Situs](https://www.sigmaaldrich.com/ID/id/site-map)
An unknown error has occured.