Skip to Content
Merck

197334

InSolution Bcl-2 Inhibitor V 1PC X 5MG

Synonym(s):

4-(4-((4ʹ-Chlorobiphenyl-2-yl)methyl)piperazin-1-yl)-N-((4-(((1R)-3-(dimethylamino)-1-((phenylsulfanyl)methyl)propyl)amino)-3-nitrophenyl)sulfonyl)benzamide, (R)-4-(4-((4ʹ-Chlorobiphenyl-2-yl)methyl)piperazin-1-yl)-N-(4-(4-(dimethylamino)-1-(phenylthio), InSolution Bcl-2 Inhibitor V 1PC X 5MG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C42H45ClN6O5S2
UNSPSC Code:
12352202
NACRES:
NA.28
Form:
liquid
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Level

storage temp.

−70°C

General description

A cell-permeable nitrophenylsulfonylbenzamide compound that is shown to bind anti-apoptotic Bcl-2 family members Bcl-2, Bcl-XL, and Bcl-w with high affinity (Ki ≤1 nM) and serves as a nonpeptidyl alternative to Bad-derived BH3 peptide (Cat. No. 197220) in preventing the binding and inhibition of the pro- apoptotic by the anti-apoptotic Bcl-2 family proteins. ABT-737 is demonstrated to fully reverse the blockage of Myr-Bid-induced cytochrome c release by Bcl-2 in mitochondria preparations from Bcl-2 overexpressing FL5.12 cells and effectively inhibit the growth of numerous cancer cells both in cultures in vitro (IC50 ≤100 nM against H146 and H1963) and in vivo (complete regression of H146 and H1963 in xenograft mice at 100 mg/kg/day, i.p.). Application of ABT-737 is also demonstrated to significantly reduce the disease severity in several murine models of autoimmunity.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
197334-5MG04055977206111