Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C16H11N3OS
CAS Number:
Molecular Weight:
293.34
UNSPSC Code:
12352200
Assay
≥93% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
off-white
solubility
DMSO: 100 mg/mL
shipped in
wet ice
storage temp.
−20°C
InChI
1S/C16H11N3OS/c21-16-18-17-15(19(16)12-7-2-1-3-8-12)14-10-11-6-4-5-9-13(11)20-14/h1-10H,(H,18,21)
InChI key
CPQLCYNKRBXXNA-UHFFFAOYSA-N
General description
A triazole that is shown to activate Caspase-3 (EC50 = 50 µM) from HeLa membrane extracts, by relieving the inhibitory effects of ProT during apoptosome formation.
Warning
Toxicity: Standard Handling (A)
Other Notes
Xin, Q., et al. 2009. Biochem.9, 1923.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service