99100106
Opentrons® 1000μL tip rack, filtered
Tips for AAW™ Workstation
Select a Size
About This Item
PCR clean
RNase and DNase free
autoclavable
endotoxin free
pyrogen free
Skip To
material
polypropylene
sterility
sterile
feature
DNA free (Human)
PCR clean
RNase and DNase free
autoclavable
endotoxin free
pyrogen free
manufacturer/tradename
991-00106
packaging
pkg of 20 packs of 96 each
volume
1000 μL
Related Categories
Related Content
View an automated protocol for Duolink® assays on the AAW™ automated assay workstation and see results comparing manual vs automated runs.
Read an automated protocol for protein purification using PureProteome™ nickel magnetic beads on the AAW™ automated assay workstation and see results comparing manual vs automated runs.
Read an automated protocol for SMC® immunoassays on the AAW™ automated assay workstation and compare manual vs automated runs.