Skip to Content
Products
Cart
0
IE
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
4-PHENYL-HEXAN-3-OL
Linear Formula:
C
12
H
18
O
CAS Number:
6006-69-5
Molecular Weight:
178.277
Products
Special Grade
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
S494003
4-PHENYL-HEXAN-3-OL,
Aldrich
CPR
Expand
View Pricing
S494607
4-PHENYL-HEXAN-3-OL,
Aldrich
CPR
Expand
View Pricing
S316024
4-PHENYL-HEXAN-3-OL,
Aldrich
CPR
Expand
View Pricing
S332879
4-PHENYL-HEXAN-3-OL,
Aldrich
CPR
Expand
View Pricing
An unknown error has occured.