All Photos(1)
Z740623
Sigma-Aldrich
Aldrich® Essentials crystallizing dish, without spout
capacity 45 mL, dish diam. × H 40 mm × 25 mm
NACRES:
NB.43
material
borosilicate glass
packaging
pack of 24 ea
capacity
45 mL
dish diam. × H
40 mm × 25 mm
General description
Clear borosilicate glass crystalizing dishes, with pour spout for easy dispensing of liquids. Not graduated.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).