Skip to Content
Merck
Hybridization solution

Hybridization solution

Hybridization solution
Synonym(s):
Hyb solution, Hybridization solution for in situ hybridization, Ready-to-use hybridization solution
EC Number:

Products

Product Category
Brand
Physical Form
Quality Segment
Technique
Showing 1-2 of 2