IL
EN
Applications
Products
Services
Support
Home
Search Results
OmniPur
®
Water, WFI Quality, Sterile Purified Water, Cell Culture Tested
(1)
OmniPur
®
Water, WFI Quality, Sterile Purified Water, Cell Culture Tested
Synonyms:
Water
Linear Formula:
H
2
O
CAS Number:
7732-18-5
Molecular Weight:
18.02
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
4.86505
OmniPur
®
Water, WFI Quality, Sterile Purified Water, Cell Culture Tested
Expand
View Pricing