Merck
  • An anaerobic continuous-flow fixed-bed reactor sustaining a 3-chlorobenzoate-degrading denitrifying population utilizing versatile electron donors and acceptors.
An anaerobic continuous-flow fixed-bed reactor sustaining a 3-chlorobenzoate-degrading denitrifying population utilizing versatile electron donors and acceptors.
Chemosphere (2004-01-15)
Hee-Sung Bae, Takao Yamagishi, Yuichi Suwa
ABSTRACT
An anaerobic continuous-flow fixed-bed column reactor capable of degrading 3-chlorobenzoate (3-CBA) under denitrifying conditions was established, and its rate reached 2.26 mM d(-1). The denitrifying population completely degraded 3-CBA when supplied at 0.1-0.54 mM, but its activity was partly suppressed when 3-CBA was supplied at 0.89 mM. Nitrate was concomitantly consumed throughout the operation of the reactor, the amount of which was similar to or up to 35% higher than the theoretical stoichiometric value that was calculated by assuming that 3-CBA degradation is coupled with denitrification. Batch incubation experiments proved that nitrate is strictly required for 3-CBA degradation in the absence of molecular oxygen. The population also degraded 3-CBA aerobically. Benzoate and 4-CBA were degraded under denitrifying conditions as well as 3-CBA, but 2-CBA was not. Considering that the previously reported denitrifying 3-CBA-degrading cultures do not exhibit 4-CBA degradation under denitrifying conditions, nor aerobic 3-CBA degradation [FEMS Microbiol. Lett. 144 (1996) 213, Appl. Environ. Microbiol. 66 (2000) 3446], the microbial population developed in this experiment was physiologically versatile with respect to the utilization of both electron donors and electron acceptors.
Sigma-Aldrich
3-Chlorobenzoic acid, ReagentPlus®, ≥99%
Pricing and availability is not currently available.