Skip to Content
Merck
All Photos(1)

Key Documents

CLS352001

Corning® Falcon test tube, with snap cap

capacity 14 mL, round polystyrene bottom, individually wrapped, sterile

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
₹10,356.85

₹10,356.85


Please contact Customer Service for Availability


Select a Size

Change View
500 EA
₹10,356.85

About This Item

UNSPSC Code:
41121800

₹10,356.85


Please contact Customer Service for Availability

material

round polystyrene bottom

sterility

sterile

feature

graduations: no
individually wrapped

packaging

pack of 500 ea

manufacturer/tradename

Corning 352001

capacity

14 mL

tube diam.

17 mm , approx.

color

clear

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service