Skip to Content
Merck

CIAH987F08B5

ChemScene LLC (Preferred Partner)

Fmoc-Lys(Me)2-OH (hydrochloride)

Synonym(s):

(2S)-6-(dimethylamino)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

100 MG

₹12,759.50

250 MG

₹14,421.25

1 G

₹19,993.00

5 G

₹55,965.00

10 G

₹1,00,832.25

25 G

₹1,89,296.00

Vendor SKU: CS-0005660-100MG

₹12,759.50


Ships in 12 days - from Aldrich Partner.


About This Item

Linear Formula:
C23H29O4N2Cl1
CAS Number:
Molecular Weight:
432.95
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C23H28N2O4.ClH/c1-25(2)14-8-7-13-21(22(26)27)24-23(28)29-15-20-18-11-5-3-9-16(18)17-10-4-6-12-19(17)20;/h3-6,9-12,20-21H,7-8,13-15H2,1-2H3,(H,24,28)(H,26,27);1H/t21-;/m0./s1

Inchi Key

SJFAFKDBWNFBCC-BOXHHOBZSA-N

Purity

97%

Salt Data

Hydrochloride

Storage temp.

4C, stored under nitrogen

Country of Origin

CN

Physical Form

Solid

Melting Point

>107 C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service