Skip to Content
Merck

PAP10

Anti-PAP-Goat Antibody

Sign In to View Organizational & Contract Pricing

Select a Size

250 μL

₹15,830.00

₹15,830.00


Estimated to ship on18 December 2025Details


Request a Bulk Order

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.77

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

antibody form

purified antibody

Quality Level

clone

polyclonal

species reactivity

goat

concentration

20 mg/mL

technique(s)

immunohistochemistry: suitable

General description

PAP soluble complexes are recommended when high sensitivity immunological detection of antigens or primary antibodies is required.

Whole antisera against IgG are suggested for bridging PAP to primary antibodies. Anti-whole IgG or anti-F(ab′)2 fragments of IgG will also bridge PAP to IgM primary antibodies, by virtue of common light chain recognition. Normal serums from the same host species as the bridging antibodies are suggested as blocking agents to minimize non-specific binding.

Biochem/physiol Actions

SOLUBLE COMPLEX, Goat PAP

Physical form

Lyophilized. Buffer= 0.01M Sodium phosphate, 0.25 M NaCl containing 0.01% thimerosal, pH 7.6.

RECONSTITUTION: Reconstitute to proper volume with sterile distilled water. Let stand 1-2 hours at room temperature.

Preparation Note

Maintain lyophilized material at 2-8°C for up to 12 months. After reconstitution, aliquot and freeze at -20°C in convenient undiluted aliquots for up to 6 months. WARNING: The use of sodium azide as a preservative will substantially inhibit the enzyme activity of HRP.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service