Merck
All Photos(1)

81277

Sigma-Aldrich

Polycaprolactone

Synonym(s):
6-Caprolactone polymer, 2-Oxepanone homopolymer
Linear Formula:
(C6H10O2)n

mp

60 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Biodegradable polymer
Non-toxic, biodegradable in soil, broad miscibility, mechanical compatibility with many polymers and good adhesion to a broad spectrum of substrates.

Other Notes

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service