Skip to Content
Products
Cart
0
IN
EN
Products
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Fmoc-Lys(Me)2-OH (hydrochloride)
CAS Number:
252049-10-8
Molecular Weight:
432.95
Products
Brand
ChemScene LLC (Preferred Partner)
ChemScene LLC / Jinay Pharmaceuticals Pvt Ltd. (Preferred Partner)
Purity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
CIAH987F08B5
Fmoc-Lys(Me)2-OH (hydrochloride),
Expand
View Pricing
JAYH9A8E9665
Fmoc-Lys(Me)2-OH (hydrochloride),
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Learn More