Skip to Content
Products
Cart
0
IN
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
ULK2, active
Products
Brand
Sigma-Aldrich
Biological Source
Quality Segment
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
14-772-M
ULK2, active,
Expand
View Pricing
An unknown error has occured.