- Selective A
Selective A
Metabolic brain disease (2017-08-11)
Cristiani F Bortolatto, Angélica S Reis, Mikaela P Pinz, Guilherme T Voss, Renata L Oliveira, Ane G Vogt, Silvane Roman, Cristiano R Jesse, Cristiane Luchese, Ethel A Wilhelm
PMID28795281
ABSTRACT
The aim of the present study was to investigate the effects of SCH58261, a selective adenosine A