Merck
GC Analysis of Column Bleed on SLB®-IL60 using FID, Comparison to 5 Polyethylene Glycol (PEG) Columns
GC Analysis of Column Bleed on SLB®-IL60 using FID, Comparison to 5 Polyethylene Glycol (PEG) Columns suitable for GC
Materials
related product
Product No.
Description
Pricing
SLB®-IL60 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
30 m x 0.25 mm I.D., 0.25 μm
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
oven
50 °C (2 min), 15 °C/min to column programmed temperature limit (10
inj. temp.
250 °C
detector
FID, at column programmed temperature limit
carrier gas
helium, 1 mL/min
injection
1.0 μL splitless
liner
4 mm I.D., split/splitless type, single taper wool packed FocusLiner design
sample
methylene chloride
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany