Merck
HPLC Analysis of Horny Goat Weed Extract on Ascentis® Express C18
HPLC Analysis of Horny Goat Weed Extract on Ascentis® Express C18 application for HPLC
Materials
related product
Product No.
Description
Pricing
Ascentis® Express C18, 5 μm HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
sample preparation
to 100 mg of Horny Goat Weed herbal supplement add 10 mL ethanol, sonicate for 30 min, centrifuge 10,000 rpm for 1 min, filter supernatant using 0.45 μm filter, dilute to 30:70, water:ethanol
column
Ascentis Express C18, 15 cm x 4.6 mm I.D., 5 μm particles (50537-U)
mobile phase
(A) 0.1% acetic acid in water; (B) 0.1% acetic acid in acetonitrile;
gradient
5% B for 0.75 min; to 40% B in 3.75 min; to 100% B in 3.5 min; held at 100% B for 2 min
flow rate
1.5 mL/min
pressure
2120 psi (146 bar)
column temp.
35 °C
detector
UV, 250 nm
injection
5 μL
sample
each compound, 50 μg/mL in water:methanol (70:30)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany