99900196
AAW™ Pipettes
Opentrons Flex™ 96-Channel Pipette
Select a Size
1 EACH
€13,730.00
€13,730.00
Please contact Customer Service for Availability
Skip To
feature
Smart sensors
packaging
pack of 1 ea
manufacturer/tradename
999-00196
capacity
5—1000 μL
Related Categories
General description
The OPENTRONSFLEX™ 96-Channel pipette enables accurate high throughput pipetting, optimized for working in 96-well plates. Flex Pipettes use air displacement technology to offer highly accurate pipetting with pipette volume ranges from 5 – 1000 μL.Smart sensors support automatic calibration, real-time positioning, and error detection. The 96-channel pipette occupies both pipette mounts and can easily be removed and swapped to an alternate pipette configurations to adapt your workflow needs.Four 96-Channel Tip Rack Adapters are included with the purchase of one Flex 96-Channel Pipette.
Legal Information
AAW is a trademark of Millipore Corporation
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.