Skip to Content
Products
Cart
0
IT
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Monoclonal Anti-CD3 PerCP produced in mouse
Synonym(s):
CD3E, T3E, TCRE
Products
Product Category
primary antibodies
Clonality
Physical Form
Conjugate
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SAB4702135
Monoclonal Anti-CD3 PerCP produced in mouse,
Expand
View Pricing
SAB4702148
Monoclonal Anti-CD3 PerCP produced in mouse,
Expand
View Pricing
An unknown error has occured.