Merck
  • Pregnancy-associated diamine oxidase originates from extravillous trophoblasts and is decreased in early-onset preeclampsia.
Pregnancy-associated diamine oxidase originates from extravillous trophoblasts and is decreased in early-onset preeclampsia.
Scientific reports (2018-04-22)
Philipp Velicky, Karin Windsperger, Karin Petroczi, Sophie Pils, Birgit Reiter, Tamara Weiss, Sigrid Vondra, Robin Ristl, Sabine Dekan, Christian Fiala, David E Cantonwine, Thomas F McElrath, Bernd Jilma, Martin Knöfler, Thomas Boehm, Jürgen Pollheimer
ABSTRACT
Human extravillous trophoblast (EVT) invasion of the pregnant uterus constitutes a pivotal event for the establishment of the maternal-fetal interface. Compromised EVT function manifesting in inadequate arterial remodeling is associated with the severe pregnancy disorder early-onset preeclampsia (eoPE). Recent studies suggest that EVTs invade the entire uterine vasculature including arteries, veins and lymphatics in the first trimester of pregnancy. We therefore hypothesized that EVT-derived factors accumulate in the circulation of pregnant women early in gestation and may serve to predict eoPE. In contrast to published literature, we demonstrate that placenta-associated diamine oxidase (DAO) is not expressed by maternal decidual cells but solely by EVTs, especially when in close proximity to decidual vessels. Cultures of primary EVTs express and secret large amounts of bioactive DAO. ELISA measurements indicate a pregnancy-specific rise in maternal DAO plasma levels around gestational week (GW) 7 coinciding with vascular invasion of EVTs. Strikingly, DAO levels from eoPE cases were significantly lower (40%) compared to controls in the first trimester of pregnancy but revealed no difference at mid gestation. Furthermore, DAO-containing pregnancy plasma rapidly inactivates pathophysiologically relevant histamine levels. This study represents the first proof of concept suggesting EVT-specific signatures as diagnostic targets for the prediction of eoPE.
Product Description
Supelco
Diminazene aceturate, analytical standard
Sigma-Aldrich
Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Carbonate-Bicarbonate Buffer, capsule
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross adsorbed-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder