Merck
  • Mitochondrial NADP+-dependent isocitrate dehydrogenase deficiency increases cisplatin-induced oxidative damage in the kidney tubule cells.
Mitochondrial NADP+-dependent isocitrate dehydrogenase deficiency increases cisplatin-induced oxidative damage in the kidney tubule cells.
Cell death & disease (2018-04-27)
Min Jung Kong, Sang Jun Han, Jee In Kim, Jeen-Woo Park, Kwon Moo Park
ABSTRACT
Mitochondrial NADP+-dependent isocitrate dehydrogenase (IDH2) plays an important role in the formation of NADPH, which is critical for the maintenance of mitochondrial redox balance. Cis-diamminedichloroplatinum II (cisplatin), an effective anticancer drug, induces oxidative stress-related nephrotoxicity, limiting its use. Therefore, we investigated whether IDH2, which is a critical enzyme in the NADPH-associated mitochondrial antioxidant system, is involved in cisplatin nephrotoxicity. Idh2 gene-deleted (Idh2-/-) mice and wild-type (Idh2 +/+ ) littermates were treated with cisplatin, with or without 2-(2,2,6,6-tetramethylpiperidin-1-oxyl-4-ylamino)-2-oxoethyl) triphenylphosphonium chloride (Mito-T), a mitochondria-specific antioxidant. Cisplatin-induced renal functional and morphological impairments were greater in Idh2-/- mice than in Idh2 +/+ mice. Mito-T mitigated those impairments in both Idh2-/- and Idh2 +/+ mice and this mitigation was greater in Idh2-/- than in Idh2 +/+ mice. Cisplatin impaired IDH2 function in the mitochondria, decreasing mitochondrial NADPH and GSH levels and increasing H2O2 generation; protein, lipid, and DNA oxidation; mitochondrial damage; and apoptosis. These cisplatin-induced changes were much more severe in Idh2-/- mice than in Idh2 +/+ mice. Mito-T treatment attenuated cisplatin-induced alterations in both Idh2-/- and Idh2 +/+ mice and this mitigation was greater in Idh2-/- than in Idh2 +/+ mice. Altogether, these data demonstrate that cisplatin induces the impairment of the mitochondrial IDH2-NADPH-GSH antioxidant system and IDH2 deficiency aggravates cisplatin-induced mitochondrial oxidative damage, inducing more severe nephrotoxicity. This suggests that the mitochondrial IDH2-NADPH-GSH antioxidant system is a target for the prevention of cisplatin-induced kidney cell death.